Gene Transcripts

Gene Symbol
ADAM3A
Entrez Gene ID
1587
Gene Name
ADAM metallopeptidase domain 3A (pseudogene)
Gene Aliases
ADAM3, CYRN1, tMDCI
Chromosome Location
Chr. 8: 39451045 - 39522989 on Build GRCh38
UniGene ID
Hs.98848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001569.3 15 1782
NR_024106.2 16 1877
NR_024107.2 17 2028
NR_073423.1 17 2125

View Full Product Details