Gene Transcripts

Gene Symbol
SFTA1P
Entrez Gene ID
207107
Gene Name
surfactant associated 1, pseudogene
Gene Aliases
SFTPF
Chromosome Location
Chr. 10: 10784439 - 10794914 on Build GRCh38
UniGene ID
Hs.31562
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027082.1 1 57
GenBank mRNA AY102069.1 - 1 57


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000446372.2 - 1 57 Chr. 10 - 10784439 - 10794914 [-]

View Full Product Details