Gene Transcripts

Gene Symbol
ANKRD30BP2
Entrez Gene ID
149992
Gene Name
ankyrin repeat domain 30B pseudogene 2
Gene Aliases
C21orf99, CT85, CTSP-1, CTSP1
Chromosome Location
Chr. 21: 13038166 - 13118251 on Build GRCh38
UniGene ID
Hs.558645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026916.1 7, 8 2046
GenBank mRNA DQ179139.1 - 1, 2 114

View Full Product Details