Gene Transcripts

Gene Symbol
LINC01550
Entrez Gene ID
388011
Gene Name
long intergenic non-protein coding RNA 1550
Gene Aliases
C14orf64
Chromosome Location
Chr. 14: 97925610 - 97978124 on Build GRCh38
UniGene ID
Hs.651477
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015430.1 2 208
GenBank mRNA AK091668.1 - 2 194
BC043585.1 - 2 208
BX647564.1 - 2 192
JX073283.1 - 2 144


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA521036.1 - 2 194 Chr. 14 - 97960263 - 97978110 [-]
GenBankmRNA BX109572.1 - 2 153 Chr. 14 - 97968300 - 97978070 [-]
GenBankmRNA DB154307.1 - 2 149 Chr. 14 - 97968842 - 97978065 [-]
GenBankmRNA BI834723.1 - 2 211 Chr. 14 - 97926352 - 97978124 [-]
GenBankmRNA AA418794.1 - 2 121 Chr. 14 - 97968315 - 97978037 [-]

View Full Product Details