Gene Transcripts
- Gene Symbol
- NKAPP1
- Entrez Gene ID
- 158801
- Gene Name
- NFKB activating protein pseudogene 1
- Gene Aliases
- CXorf42
- Chromosome Location
- Chr. X: 120236452 - 120245267 on Build GRCh38
- UniGene ID
- Hs.442518
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027131.1 |
|
3 |
2149 |
|
GenBank mRNA |
AK093895.1 |
- |
3 |
2115 |
|
BC022359.1 |
- |
3 |
2089 |
|
View Full Product Details