Gene Transcripts
- Gene Symbol
- TDRG1
- Entrez Gene ID
- 732253
- Gene Name
- testis development related 1 (non-protein coding)
- Gene Aliases
- LINC00532
- Chromosome Location
- Chr. 6: 40378424 - 40379892 on Build GRCh38
- UniGene ID
- Hs.180197
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000448559.5 |
- |
2 |
560 |
|
Chr. 6 - 40340465 - 40379885 [+] |
GenBankmRNA |
AL045589.1 |
- |
2 |
691 |
|
Chr. 6 - 40378663 - 40379717 [+] |
GENCODE |
ENST00000451810.2 |
- |
3 |
736 |
|
Chr. 6 - 40334954 - 40379885 [+] |
GENCODE |
ENST00000373170.2 |
- |
2 |
984 |
|
Chr. 6 - 40378337 - 40379899 [+] |
View Full Product Details