Gene Transcripts
- Gene Symbol
- ST7-AS1
- Entrez Gene ID
- 93653
- Gene Name
- ST7 antisense RNA 1
- Gene Aliases
- ST7AS1, ST7OT1
- Chromosome Location
- Chr. 7: 116952446 - 116954334 on Build GRCh38
- UniGene ID
- Hs.597516
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI262731.1 |
- |
1 |
314 |
|
Chr. 7 - 116952446 - 116953370 [-] |
GenBankmRNA |
BI262735.1 |
- |
1 |
152 |
|
Chr. 7 - 116952581 - 116953207 [-] |
GenBankmRNA |
BI262730.1 |
- |
1 |
301 |
|
Chr. 7 - 116952574 - 116953356 [-] |
View Full Product Details