Gene Transcripts
- Gene Symbol
- LINC00312
- Entrez Gene ID
- 29931
- Gene Name
- long intergenic non-protein coding RNA 312
- Gene Aliases
- ERR-10, ERR10, LMCD1DN, LOH3CR2A, NAG-7, NAG7, NCRNA00312
- Chromosome Location
- Chr. 3: 8571782 - 8574668 on Build GRCh38
- UniGene ID
- Hs.433151
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA283550.1 |
- |
1 |
391 |
|
Chr. 3 - 8572088 - 8572650 [+] |
View Full Product Details