Gene Transcripts
- Gene Symbol
- ANO1
- Entrez Gene ID
- 55107
- Gene Name
- anoctamin 1
- Gene Aliases
- DOG1, ORAOV2, TAOS2, TMEM16A
- Chromosome Location
- Chr. 11: 70078302 - 70189545 on Build GRCh38
- UniGene ID
- Hs.503074
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_030691.1 |
|
13, 14 |
1565 |
|
XM_006718605.2 |
XP_006718668.1
|
13, 14 |
1560 |
|
XM_011545125.2 |
XP_011543427.1
|
15, 16 |
1503 |
|
XM_011545126.2 |
XP_011543428.1
|
14, 15 |
1491 |
|
XM_011545127.2 |
XP_011543429.1
|
14, 15 |
1437 |
|
XM_011545128.2 |
XP_011543430.1
|
13, 14 |
1425 |
|
XM_011545131.2 |
XP_011543433.1
|
13, 14 |
1433 |
|
GenBank mRNA |
AK001123.1 |
- |
5, 6 |
345 |
|
AK097619.1 |
- |
13, 14 |
1433 |
|
BC027590.2 |
- |
9, 10 |
681 |
|
BC033036.2 |
- |
13, 14 |
1521 |
4823645 |
View Full Product Details