Gene Transcripts
- Gene Symbol
- FAM215A
- Entrez Gene ID
- 23591
- Gene Name
- family with sequence similarity 215 member A (non-protein coding)
- Gene Aliases
- APR-2, C17orf88, LINC00530
- Chromosome Location
- Chr. 17: 43917208 - 43917987 on Build GRCh38
- UniGene ID
- Hs.721804
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026770.1 |
|
1 |
176 |
|
GenBank mRNA |
AF143236.1 |
- |
1 |
188 |
|
BC069097.1 |
- |
1 |
176 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AX721312.1 |
- |
1 |
1205 |
|
Chr. 17 - 43133910 - 43136073 [+] |
View Full Product Details