Gene Transcripts

Gene Symbol
TTC41P
Entrez Gene ID
253724
Gene Name
tetratricopeptide repeat domain 41, pseudogene
Gene Aliases
GNN, GNNP
Chromosome Location
Chr. 12: 103843749 - 103930211 on Build GRCh38
UniGene ID
Hs.506549
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027249.1 2 784
GenBank mRNA AK299490.1 - 2 704
BC016335.1 - 2 616
BC033128.1 - 6 1365

View Full Product Details