Gene Transcripts
- Gene Symbol
- NHSL2
- Entrez Gene ID
- 340527
- Gene Name
- NHS like 2
- Gene Aliases
- Chromosome Location
- Chr. X: 71911073 - 72153286 on Build GRCh38
- UniGene ID
- Hs.397836
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA682621.1 |
- |
1 |
401 |
|
Chr. X - 72151337 - 72151903 [+] |
GenBankmRNA |
CX752026.1 |
- |
1 |
424 |
|
Chr. X - 72151323 - 72152030 [+] |
GenBankmRNA |
BE620108.1 |
- |
1 |
313 |
|
Chr. X - 72151422 - 72152229 [+] |
View Full Product Details