Gene Transcripts
- Gene Symbol
- EVA1C
- Entrez Gene ID
- 59271
- Gene Name
- eva-1 homolog C (C. elegans)
- Gene Aliases
- B18, B19, C21orf63, C21orf64, FAM176C, PRED34, SUE21
- Chromosome Location
- Chr. 21: 32411692 - 32515400 on Build GRCh38
- UniGene ID
- Hs.208358
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126660.1 |
- |
1 |
570 |
|
View Full Product Details