Gene Transcripts
- Gene Symbol
- JOSD1
- Entrez Gene ID
- 9929
- Gene Name
- Josephin domain containing 1
- Gene Aliases
- dJ508I15.2
- Chromosome Location
- Chr. 22: 38685543 - 38701418 on Build GRCh38
- UniGene ID
- Hs.3094
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX538323.1 |
- |
|
884 |
|
View Full Product Details