Gene Transcripts

Gene Symbol
CAMTA1
Entrez Gene ID
23261
Gene Name
calmodulin binding transcription activator 1
Gene Aliases
CANPMR
Chromosome Location
Chr. 1: 6785324 - 7769706 on Build GRCh38
UniGene ID
Hs.397705
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242701.1 NP_001229630.1 4 694
NR_038934.1 4 708
GenBank mRNA AF086286.1 - 1 105
BM470492.1 - 569
CD103791.1 - 4 493


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG202170.1 - 2 361 Chr. 1 - 6886244 - 6888201 [+]
GenBankmRNA BM800184.1 - 3 468 Chr. 1 - 6825088 - 6888201 [+]
GenBankmRNA W69701.1 - 106 Chr. - - [-]
GenBankmRNA BG208453.1 - 2 341 Chr. 1 - 6871756 - 6888201 [+]
GenBankmRNA BG208979.1 - 2 341 Chr. 1 - 6871754 - 6888201 [+]
GenBankmRNA BG028550.1 - 408 Chr. - - [-]
GenBankmRNA DB636726.2 - 3 432 Chr. 1 - 6820189 - 6888199 [+]
GenBankmRNA BG206886.1 - 2 342 Chr. 1 - 6871756 - 6888201 [+]
GenBankmRNA BG184758.1 - 3 444 Chr. 1 - 6871757 - 6888186 [+]
GenBankmRNA BF448274.1 - 2 319 Chr. 1 - 6871787 - 6888141 [+]
GenBankmRNA BG218364.1 - 2 362 Chr. 1 - 6886244 - 6888201 [+]
GenBankmRNA BG185174.1 - 2 342 Chr. 1 - 6871754 - 6888201 [+]

View Full Product Details