Gene Transcripts
- Gene Symbol
- ASTN2
- Entrez Gene ID
- 23245
- Gene Name
- astrotactin 2
- Gene Aliases
- bA67K19.1
- Chromosome Location
- Chr. 9: 116425225 - 117415039 on Build GRCh38
- UniGene ID
- Hs.601562
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX648673.1 |
- |
1 |
6460 |
|
View Full Product Details