Gene Transcripts
- Gene Symbol
- ANK3
- Entrez Gene ID
- 288
- Gene Name
- ankyrin 3, node of Ranvier (ankyrin G)
- Gene Aliases
- ANKYRIN-G, MRT37
- Chromosome Location
- Chr. 10: 60026298 - 60733526 on Build GRCh38
- UniGene ID
- Hs.499725
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK172736.1 |
- |
3 |
808 |
|
View Full Product Details