Gene Transcripts
- Gene Symbol
- COLCA2
- Entrez Gene ID
- 120376
- Gene Name
- colorectal cancer associated 2
- Gene Aliases
- C11orf93, CASC13, LOH11CR1G
- Chromosome Location
- Chr. 11: 111298517 - 111308735 on Build GRCh38
- UniGene ID
- Hs.31409
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB096246.1 |
- |
1 |
1827 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM821976.1 |
- |
1 |
480 |
|
Chr. 11 - 111303379 - 111303887 [+] |
View Full Product Details