Gene Transcripts
- Gene Symbol
- TTC39C
- Entrez Gene ID
- 125488
- Gene Name
- tetratricopeptide repeat domain 39C
- Gene Aliases
- C18orf17, HsT2697
- Chromosome Location
- Chr. 18: 23992773 - 24135952 on Build GRCh38
- UniGene ID
- Hs.128576
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BM069557.1 |
- |
1 |
105 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF363068.1 |
- |
|
311 |
|
Chr. - - [-] |
GenBankmRNA |
CA774026.1 |
- |
1 |
450 |
|
Chr. 18 - 24135430 - 24135968 [-] |
View Full Product Details