Gene Transcripts

Gene Symbol
ARRDC5
Entrez Gene ID
645432
Gene Name
arrestin domain containing 5
Gene Aliases
Chromosome Location
Chr. 19: 4867614 - 4915469 on Build GRCh38
UniGene ID
Hs.574574
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011528181.2 XP_011526483.1 1 88
GenBank mRNA AK097879.1 - 1 1438

View Full Product Details