Gene Transcripts

Gene Symbol
AJAP1
Entrez Gene ID
55966
Gene Name
adherens junctions associated protein 1
Gene Aliases
MOT8, SHREW-1, SHREW1
Chromosome Location
Chr. 1: 4654775 - 4792518 on Build GRCh38
UniGene ID
Hs.12473
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011541786.2 XP_011540088.1 7 11804
GenBank mRNA BC037321.2 - 1 2614

View Full Product Details