Gene Transcripts
- Gene Symbol
- METTL15
- Entrez Gene ID
- 196074
- Gene Name
- methyltransferase like 15
- Gene Aliases
- METT5D1
- Chromosome Location
- Chr. 11: 28108251 - 28527041 on Build GRCh38
- UniGene ID
- Hs.243326
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ131606.1 |
- |
1 |
190 |
|
View Full Product Details