Gene Transcripts

Gene Symbol
ELP6
Entrez Gene ID
54859
Gene Name
elongator acetyltransferase complex subunit 6
Gene Aliases
C3orf75, TMEM103
Chromosome Location
Chr. 3: 47495640 - 47513711 on Build GRCh38
UniGene ID
Hs.311100
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011533851.2 XP_011532153.1 5 723
XM_011533852.2 XP_011532154.1 5 1059
XM_011533853.2 XP_011532155.1 5 700
XM_011533854.2 XP_011532156.1 4 624
XM_011533856.2 XP_011532158.1 4 679
XM_011533857.2 XP_011532159.1 4 849


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU566657.1 - 1 151 Chr. 3 - 47501438 - 47501723 [-]
GenBankmRNA CR619601.1 - 1 138 Chr. 3 - 47501440 - 47501710 [-]
GenBankmRNA BX403218.2 - 1 138 Chr. 3 - 47501440 - 47501710 [-]
GenBankmRNA BX341318.2 - 1 138 Chr. 3 - 47501440 - 47501710 [-]
GenBankmRNA CB306887.1 - 5 602 Chr. 3 - 47501527 - 47513025 [-]

View Full Product Details