Gene Transcripts

Gene Symbol
Mxd4
Entrez Gene ID
17122
Gene Name
Max dimerization protein 4
Gene Aliases
2810410A03Rik, Mad4, bHLHc12
Chromosome Location
Chr. 5: 34173887 - 34187750 on Build GRCm38
UniGene ID
Mm.259260
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK013062.1 - 1 697
BC025016.1 - 1 932


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BE374677.1 - 1 398 Chr. 5 - 34173887 - 34174393 [-]
GenBankmRNA BF720640.1 - 2 405 Chr. 5 - 34173887 - 34174887 [-]
GenBankmRNA BY712114.1 - 1 698 Chr. 5 - 34173887 - 34174691 [-]
GenBankmRNA BU614560.1 - 1 490 Chr. 5 - 34173887 - 34174485 [-]
GenBankmRNA BB830811.1 - 1 330 Chr. 5 - 34173888 - 34174325 [-]
GenBankmRNA BI736434.1 - 1 614 Chr. 5 - 34173896 - 34174609 [-]
GenBankmRNA BB771636.1 - 1 429 Chr. 5 - 34173888 - 34174426 [-]
GenBankmRNA AA435430.1 - 1 345 Chr. 5 - 34173910 - 34174336 [-]
GenBankmRNA AA016917.1 - 1 431 Chr. 5 - 34173984 - 34174426 [-]
GenBankmRNA BB835420.1 - 1 332 Chr. 5 - 34173888 - 34174327 [-]
GenBankmRNA CB193583.1 - 2 674 Chr. 5 - 34173883 - 34175336 [-]
GenBankmRNA BG868949.1 - 1 603 Chr. 5 - 34173892 - 34174596 [-]
GenBankmRNA BI146403.1 - 1 285 Chr. 5 - 34173887 - 34174280 [-]
GenBankmRNA CN531960.1 - 1 544 Chr. 5 - 34173887 - 34174539 [-]
GenBankmRNA AA197634.1 - 1 466 Chr. 5 - 34173919 - 34174463 [-]
GenBankmRNA BB744886.1 - 1 351 Chr. 5 - 34173887 - 34174347 [-]
GenBankmRNA BB835913.1 - 1 329 Chr. 5 - 34173888 - 34174318 [-]
GenBankmRNA CB205787.1 - 565 Chr. - - [-]
GenBankmRNA BU524612.1 - 369 Chr. - - [-]
GenBankmRNA BE307748.1 - 1 238 Chr. 5 - 34173893 - 34174233 [-]

View Full Product Details