Gene Transcripts

Gene Symbol
Sfpq
Entrez Gene ID
71514
Gene Name
splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated)
Gene Aliases
1110004P21Rik, 2810416M14Rik, 5730453G22Rik, 9030402K04Rik, AU021830, D4Ertd314e, Gm12940, OTTMUSG00000009329, PSF, REP1
Chromosome Location
Chr. 4: 127020560 - 127037016 on Build GRCm38
UniGene ID
Mm.257276
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK031234.1 - 499


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB620550.1 - 3 497 Chr. 4 - 127035683 - 127048295 [+]

View Full Product Details