Gene Transcripts
- Gene Symbol
- A930005C09Rik
- Entrez Gene ID
- 77937
- Gene Name
- RIKEN cDNA A930005C09 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.17604
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020809.1 |
- |
1 |
30 |
|
- Gene Symbol
- Gnb5
- Entrez Gene ID
- 14697
- Gene Name
- guanine nucleotide binding protein (G protein), beta 5
- Gene Aliases
- GBS, Gbeta5, flr
- Chromosome Location
- Chr. 9: 75306288 - 75345923 on Build GRCm38
- UniGene ID
- Mm.17604
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CK621143.1 |
- |
1 |
75 |
|
View Full Product Details