Gene Transcripts
- Gene Symbol
- CACTIN-AS1
- Entrez Gene ID
- 404665
- Gene Name
- CACTIN antisense RNA 1
- Gene Aliases
- C19orf29-AS1, C19orf29OS
- Chromosome Location
- Chr. 19: 3607247 - 3613930 on Build GRCh38
- UniGene ID
- Hs.719726
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038865.1 |
|
4 |
1889 |
|
GenBank mRNA |
AK097999.1 |
- |
4 |
1890 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000592274.1 |
- |
4 |
1889 |
|
Chr. 19 - 3607247 - 3613930 [+] |
View Full Product Details