Gene Transcripts

Gene Symbol
PAXBP1-AS1
Entrez Gene ID
100506215
Gene Name
PAXBP1 antisense RNA 1
Gene Aliases
GCFC1-AS1
Chromosome Location
Chr. 21: 32728114 - 32743122 on Build GRCh38
UniGene ID
Hs.657123
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_038879.1 1 628
NR_038880.1 1 628
GenBank mRNA AK056384.1 - 1 627
BC038361.1 - 1 603


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000458479.1 - 1 627 Chr. 21 - 32728115 - 32732916 [+]
GENCODE ENST00000440052.5 - 1 603 Chr. 21 - 32728139 - 32743122 [+]

View Full Product Details