Gene Transcripts
- Gene Symbol
- SNCA-AS1
- Entrez Gene ID
- 644248
- Gene Name
- SNCA antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 4: 89836401 - 89841991 on Build GRCh38
- UniGene ID
- Hs.553954
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123890.1 |
- |
2 |
1733 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000501215.1 |
- |
2 |
1730 |
|
Chr. 4 - 89836424 - 89840595 [+] |
View Full Product Details