Gene Transcripts
- Gene Symbol
- TBC1D3P1-DHX40P1
- Entrez Gene ID
- 653645
- Gene Name
- TBC1D3P1-DHX40P1 readthrough, transcribed pseudogene
- Gene Aliases
- DHX40P, DHX40P1, TBC1D3P1
- Chromosome Location
- Chr. 17: 59962363 - 60019052 on Build GRCh38
- UniGene ID
- Hs.448884
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002924.3 |
|
5, 6 |
492 |
|
GenBank mRNA |
BX280208.1 |
- |
5, 6 |
415 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI828567.1 |
- |
5, 6 |
415 |
|
Chr. 17 - 59995951 - 60018975 [-] |
GENCODE |
ENST00000587125.1 |
- |
5, 6 |
415 |
|
Chr. 17 - 59962363 - 60018975 [-] |
View Full Product Details