Gene Transcripts

Gene Symbol
TBC1D3P1-DHX40P1
Entrez Gene ID
653645
Gene Name
TBC1D3P1-DHX40P1 readthrough, transcribed pseudogene
Gene Aliases
DHX40P, DHX40P1, TBC1D3P1
Chromosome Location
Chr. 17: 59962363 - 60019052 on Build GRCh38
UniGene ID
Hs.448884
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002924.3 2, 3 182
GenBank mRNA BX280208.1 - 2, 3 105


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI828567.1 - 2, 3 105 Chr. 17 - 59995951 - 60018975 [-]
GENCODE ENST00000587125.1 - 2, 3 105 Chr. 17 - 59962363 - 60018975 [-]

View Full Product Details