Gene Transcripts
- Gene Symbol
- Gm12300
- Entrez Gene ID
- 102633540
- Gene Name
- predicted gene 12300
- Gene Aliases
- OTTMUSG00000005925
- Chromosome Location
- Chr. 11: 67224194 - 67237573 on Build GRCm38
- UniGene ID
- Mm.477065
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK009488.1 |
- |
|
561 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY709201.1 |
- |
4 |
559 |
|
Chr. 11 - 67211258 - 67237573 [-] |
View Full Product Details