Gene Transcripts
- Gene Symbol
- Daf2
- Entrez Gene ID
- 13137
- Gene Name
- decay accelerating factor 2
- Gene Aliases
- AI323748, Cd55b, Daf, Daf-TM, TM-DAF
- Chromosome Location
- Chr. 1: 130388528 - 130423037 on Build GRCm38
- UniGene ID
- Mm.436759
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK019553.1 |
- |
|
1974 |
|
View Full Product Details