Gene Transcripts

Gene Symbol
Gpank1
Entrez Gene ID
81845
Gene Name
G patch domain and ankyrin repeats 1
Gene Aliases
Bat-4, Bat4, D17H6S54E, G5
Chromosome Location
Chr. 17: 35121473 - 35124815 on Build GRCm38
UniGene ID
Mm.7405
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK009290.1 - 546


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY709079.1 - 2 549 Chr. 17 - 35122751 - 35126433 [-]

View Full Product Details