Gene Transcripts

Gene Symbol
Afg3l1
Entrez Gene ID
114896
Gene Name
AFG3-like AAA ATPase 1
Gene Aliases
1700047G05Rik, 3110061K15Rik
Chromosome Location
Chr. 8: 123477862 - 123503916 on Build GRCm38
UniGene ID
Mm.287475
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054070.3 NP_473411.2 11 1391
GenBank mRNA AF329695.1 - 11 1332
AK006708.1 - 5 682
AK159647.1 - 11 1355
AK167964.1 - 11 1366
AK168244.1 - 11 1365
BC043320.1 - 2 213
BC056978.1 - 11 1350 6821746
BC060064.1 - 11 1350

View Full Product Details