Gene Transcripts

Gene Symbol
Stk33
Entrez Gene ID
117229
Gene Name
serine/threonine kinase 33
Gene Aliases
4921505G21Rik, AW061264
Chromosome Location
Chr. 7: 109279213 - 109439053 on Build GRCm38
UniGene ID
Mm.389950
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054103.1 NP_473444.1 5 878
GenBank mRNA AK014819.1 - 5 878
AM056057.1 - 2 146
BC117798.1 - 2 167
BC117799.1 - 2 167
BC127158.1 - 2 167

View Full Product Details