Gene Transcripts
- Gene Symbol
- Vmn1r22
- Entrez Gene ID
- 171196
- Gene Name
- vomeronasal 1 receptor 22
- Gene Aliases
- V1rc23
- Chromosome Location
- Chr. 6: 57900082 - 57900990 on Build GRCm38
- UniGene ID
- Mm.377175
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134178.1 |
NP_598939.1
|
1 |
517 |
|
- Gene Symbol
- Vmn1r23
- Entrez Gene ID
- 171197
- Gene Name
- vomeronasal 1 receptor 23
- Gene Aliases
- V1rc24
- Chromosome Location
- Chr. 6: 57925883 - 57926791 on Build GRCm38
- UniGene ID
- Mm.377176
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134179.1 |
NP_598940.1
|
1 |
517 |
|
View Full Product Details