Gene Transcripts
- Gene Symbol
- CDK17
- Entrez Gene ID
- 5128
- Gene Name
- cyclin dependent kinase 17
- Gene Aliases
- PCTAIRE2, PCTK2
- Chromosome Location
- Chr. 12: 96278261 - 96400588 on Build GRCh38
- UniGene ID
- Hs.506415
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001170464.2 |
NP_001163935.1
|
4, 5 |
1044 |
|
NM_002595.4 |
NP_002586.2
|
4, 5 |
1044 |
|
XM_017019405.1 |
XP_016874894.1
|
4, 5 |
486 |
|
XM_017019406.1 |
XP_016874895.1
|
4, 5 |
501 |
|
XM_017019407.1 |
XP_016874896.1
|
4, 5 |
931 |
|
GenBank mRNA |
AK290011.1 |
- |
4, 5 |
936 |
|
AK294698.1 |
- |
3, 4 |
383 |
|
AK315214.1 |
- |
4, 5 |
954 |
|
BC033005.1 |
- |
4, 5 |
533 |
4824359 |
X66360.1 |
- |
4, 5 |
481 |
|
View Full Product Details