Gene Transcripts

Gene Symbol
Best2
Entrez Gene ID
212989
Gene Name
bestrophin 2
Gene Aliases
Vmd2l1
Chromosome Location
Chr. 8: 85007202 - 85014531 on Build GRCm38
UniGene ID
Mm.215154
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130194.1 NP_001123666.1 6 771
GenBank mRNA AY450428.1 - 6 1013
BC019528.1 - 5 745 4208860
BC031186.1 - 5 732 4989959
BC036157.1 - 6 771 4982413
BC036163.1 - 6 812 4984410

View Full Product Details