Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173385.2 | NP_775561.1 | 10 | 3514 | |
GenBank mRNA | AK036473.1 | - | 10 | 3513 | |
AK081544.1 | - | 10 | 3515 | ||
BC046471.1 | - | 1 | 1542 | ||
BC058167.1 | - | 10 | 3597 | ||
BC080666.1 | - | 9 | 3510 | 4934956 |