Gene Transcripts

Gene Symbol
Aldh5a1
Entrez Gene ID
214579
Gene Name
aldhehyde dehydrogenase family 5, subfamily A1
Gene Aliases
6330403E24Rik, Ahd-1, Ahd1, D630032B01Rik, OTTMUSG00000000613, SSADH, Ssdh1
Chromosome Location
Chr. 13: 24907579 - 24937661 on Build GRCm38
UniGene ID
Mm.393311
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172532.3 NP_766120.1 2 437
GenBank mRNA AK052703.1 - 2 438
AK144030.1 - 2 421
AK159485.1 - 2 418
AK166313.1 - 2 418
BC138703.1 - 2 400 8861725

View Full Product Details