Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001115.2 | NP_001106.1 | 17 | 3482 | |
XM_005250769.3 | XP_005250826.1 | 16 | 3515 | ||
XM_006716501.3 | XP_006716564.1 | 16 | 3611 | ||
XM_017013006.1 | XP_016868495.1 | 15 | 3283 | ||
GenBank mRNA | DQ104739.1 | - | 14 | 2833 | |
GQ472223.1 | - | 17 | 5320 | ||
M83533.1 | - | 11 | 1498 | ||
Z35309.1 | - | 17 | 5320 |