Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_183154.3 | NP_898977.2 | 2 | 957 | |
GenBank mRNA | AK090334.1 | - | 3 | 1127 | |
AK129400.1 | - | 2 | 932 | ||
AK139825.1 | - | 2 | 978 | ||
AK170954.1 | - | 2 | 958 | ||
BC050015.1 | - | 1 | 378 | 30016571 |