Gene Transcripts
- Gene Symbol
- PHF1
- Entrez Gene ID
- 5252
- Gene Name
- PHD finger protein 1
- Gene Aliases
- MTF2L2, PCL1, PHF2, TDRD19C, hPHF1
- Chromosome Location
- Chr. 6: 33410751 - 33416453 on Build GRCh38
- UniGene ID
- Hs.166204
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002636.4 |
NP_002627.1
|
10, 11 |
1180 |
|
NM_024165.2 |
NP_077084.1
|
10, 11 |
1180 |
|
NR_027692.1 |
|
10, 11 |
1180 |
|
XM_011514662.1 |
XP_011512964.1
|
10, 11 |
1097 |
|
XM_011514663.1 |
XP_011512965.1
|
10, 11 |
1097 |
|
XM_011514664.1 |
XP_011512966.1
|
9, 10 |
1001 |
|
XM_011514665.1 |
XP_011512967.1
|
10, 11 |
1097 |
|
XM_011514666.1 |
XP_011512968.1
|
10, 11 |
1097 |
|
XM_011514669.1 |
XP_011512971.1
|
10, 11 |
1097 |
|
XM_011514670.2 |
XP_011512972.1
|
10, 11 |
1097 |
|
XM_017010939.1 |
XP_016866428.1
|
8, 9 |
796 |
|
GenBank mRNA |
AF029678.1 |
- |
10, 11 |
1000 |
|
AF052205.1 |
- |
10, 11 |
996 |
|
AK225913.1 |
- |
9, 10 |
1182 |
|
AK295412.1 |
- |
10, 11 |
1050 |
|
AK299162.1 |
- |
7, 8 |
679 |
|
AK316499.1 |
- |
10, 11 |
1131 |
|
BC008834.2 |
- |
10, 11 |
1148 |
4304525 |
View Full Product Details