Gene Transcripts

Gene Symbol
Bloc1s3
Entrez Gene ID
232946
Gene Name
biogenesis of lysosomal organelles complex-1, subunit 3
Gene Aliases
BC043666, Blos3, E230011O18, rp
Chromosome Location
Chr. 7: 19505804 - 19508371 on Build GRCm38
UniGene ID
Mm.64557
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177692.3 NP_808360.2 2 133
GenBank mRNA AK054006.1 - 2 152
AK153811.1 - 170
BC043666.1 - 2 133 5025443

View Full Product Details