Gene Transcripts

Gene Symbol
Pfas
Entrez Gene ID
237823
Gene Name
phosphoribosylformylglycinamidine synthase (FGAR amidotransferase)
Gene Aliases
4432409B16Rik, FGAMS, FGAR-AT, FGARAT, Gm18, PURL, Sofa
Chromosome Location
Chr. 11: 68985701 - 69008460 on Build GRCm38
UniGene ID
Mm.340288
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159519.1 NP_001152991.1 11, 12 1438
GenBank mRNA BC114997.1 - 10, 11 1372

View Full Product Details