Gene Transcripts

Gene Symbol
PSMD5
Entrez Gene ID
5711
Gene Name
proteasome 26S subunit, non-ATPase 5
Gene Aliases
S5B
Chromosome Location
Chr. 9: 120798318 - 120843021 on Build GRCh38
UniGene ID
Hs.193725
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270427.1 NP_001257356.1 7 1025
NM_005047.3 NP_005038.1 8 1154
XM_011518866.2 XP_011517168.1 8 1064
XM_017014927.1 XP_016870416.1 8 1064
XM_017014928.1 XP_016870417.1 7 954
GenBank mRNA AK026925.1 - 4 457
AK291051.1 - 8 1055
AK303007.1 - 7 955
BC014478.1 - 8 1043 4878087
D31889.1 - 8 1041
S79862.1 - 8 1045

View Full Product Details