Gene Transcripts

Gene Symbol
Hist2h2bb
Entrez Gene ID
319189
Gene Name
histone cluster 2, H2bb
Gene Aliases
H2b-616
Chromosome Location
Chr. 3: 96269700 - 96270192 on Build GRCm38
UniGene ID
Mm.297655
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175666.2 NP_783597.2 1 233
GenBank mRNA BC019122.1 - 220

View Full Product Details