Gene Transcripts

Gene Symbol
Lig4
Entrez Gene ID
319583
Gene Name
ligase IV, DNA, ATP-dependent
Gene Aliases
5830471N16Rik, tiny
Chromosome Location
Chr. 8: 9970020 - 9976323 on Build GRCm38
UniGene ID
Mm.80584
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176953.3 NP_795927.2 2 1700
GenBank mRNA AK030029.1 - 1582
AK083353.1 - 2 1580
AK148081.1 - 1676

View Full Product Details