Gene Transcripts

Gene Symbol
Cacna2d4
Entrez Gene ID
319734
Gene Name
calcium channel, voltage-dependent, alpha 2/delta subunit 4
Gene Aliases
5730412N02Rik, BE686333
Chromosome Location
Chr. 6: 119236474 - 119353392 on Build GRCm38
UniGene ID
Mm.440773
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033382.2 NP_001028554.2 21 2141
GenBank mRNA AK030723.1 - 16 1458
BC141091.1 - 20 1997 9055703

View Full Product Details